Assay Details
| Gene Symbol: Lox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017061.2 | NP_058757.1 | 4 - 5 | 1041 | 72 | |
| XM_006254713.2 | XP_006254775.1 | 2 - 3 | 1124 | 72 | ||
| XM_006254714.2 | XP_006254776.1 | 3 - 4 | 1024 | 72 | ||
| XM_006254715.2 | XP_006254777.1 | 2 - 3 | 1125 | 72 | ||
| GenBank mRNA | BC078861.1 | - | 4 - 5 | 1041 | 7113042 | 72 |
| FQ230374.1 | - | 2 - 3 | 276 | 72 | ||
| FQ231737.1 | - | 1 - 2 | 65 | 72 | ||
| U11038.1 | - | 2 - 3 | 1009 | 72 | ||
More Information
Set Membership: |
|

Lox
Genomic Map