Assay Details
| Gene Symbol: Creb3l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005562.1 | NP_001005562.1 | 5 - 6 | 1149 | 56 | |
| XM_006234584.1 | XP_006234646.1 | 5 - 6 | 1205 | 56 | ||
| GenBank mRNA | BC081951.1 | - | 5 - 6 | 1149 | 7122657 | 56 |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3l1
Genomic Map