Assay Details
| Gene Symbol: Nbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031609.1 | NP_113797.1 | 2 - 3 | 366 | 62 | |
| XM_008764284.1 | XP_008762506.1 | 2 - 3 | 330 | 62 | ||
| GenBank mRNA | BC061833.1 | - | 2 - 3 | 313 | 5599381 | 62 |
| S72637.1 | - | 2 - 3 | 366 | 62 | ||
| X66872.1 | - | 2 - 3 | 366 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nbl1
Genomic Map