Assay Details
| Gene Symbol: Mat2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044282.1 | NP_001037747.1 | 5 - 6 | 799 | 66 | |
| XM_006246126.2 | XP_006246188.1 | 5 - 6 | 949 | 66 | ||
| GenBank mRNA | BC085899.1 | - | 5 - 6 | 799 | 7193416 | 66 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mat2b
Genomic Map