Assay Details
| Gene Symbol: LOC103694875 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008773833.1 | XP_008772055.1 | 3 - 4 | 1651 | 62 | |
| XM_008773835.1 | XP_008772057.1 | 3 - 4 | 1648 | 62 | ||
| XM_008773840.1 | XP_008772062.1 | 3 - 4 | 1647 | 62 | ||
| XM_008773841.1 | XP_008772063.1 | 3 - 4 | 1648 | 62 | ||
| XM_008773842.1 | XP_008772064.1 | 3 - 4 | 1645 | 62 | ||
| Gene Symbol: Derl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109577.1 | NP_001103047.2 | 3 - 4 | 259 | 62 | |
| XM_006256325.2 | XP_006256387.1 | 3 - 4 | 1648 | 62 | ||
| XM_006256326.2 | XP_006256388.1 | 3 - 4 | 1645 | 62 | ||
| XM_008772918.1 | XP_008771140.1 | 3 - 4 | 1651 | 62 | ||
| XM_008772920.1 | XP_008771142.1 | 3 - 4 | 1648 | 62 | ||
| GenBank mRNA | BC167034.1 | - | 3 - 4 | 216 | 62 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC103694875
Genomic Map