Assay Details
| Gene Symbol: Adap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133567.2 | NP_598251.1 | 3 - 4 | 386 | 64 | |
| GenBank mRNA | AF123047.1 | - | 3 - 4 | 313 | 64 | |
| AJ007422.1 | - | 3 - 4 | 313 | 64 | ||
| AJ007616.1 | - | 3 - 4 | 313 | 64 | ||
| BC099775.1 | - | 3 - 4 | 386 | 7933317 | 64 | |
| U51013.1 | - | 3 - 4 | 413 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P323490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268225 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363713 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517380 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406595 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 340117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84778 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P323490 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P323490 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P323490 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P466159 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700496447 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1015 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22258 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700496447 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700588442 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10757107 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757116 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757105 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757106 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757111 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10757104 | - | - |
| Rat Genome 230 2.0 Array | 1368449_at | - | - |
| Rat Genome 230 2.0 Array | 1387194_at | - | - |
| Rat Genome 230 2.0 Array | 1387194_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368449_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387194_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368449_at | - | - |
| Affymetrix HT Rat Focus Array | 1368449_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387194_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200002331.rn.1 | TC1200000302.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387194_at | - | - |
| Rat Expression Set 230 | 1387194_PM_at | - | - |
| Rat Expression Set 230 | 1368449_PM_at | - | - |
| Rat Expression Set 230 | 1368449_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10757104 | - | - |
| Rat Genome U34 Set | U51013_at | - | - |
| Rat Genome U34 Set | AJ007422_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21983951 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374854 | - | - |
| RatRef-12 Expression BeadChip | ILMN_54218 | - | - |

Adap1
Genomic Map