Assay Details
| Gene Symbol: NEWGENE_2813 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762936.1 | XP_008761158.1 | 1 - 2 | 1121 | 97 | |
| Gene Symbol: Hoxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012581.2 | NP_036713.2 | 1 - 2 | 540 | 97 | |
| GenBank mRNA | M91802.1 | - | 1 - 2 | 577 | 97 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

NEWGENE_2813
Genomic Map