Assay Details
| Gene Symbol: LOC100911116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749360.3 | XP_003749408.1 | 1 - 2 | 99 | 75 | |
| Gene Symbol: Hsd3b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012584.1 | NP_036716.1 | 37 | 75 | ||
| GenBank mRNA | BC092571.1 | - | 54 | 7369915 | 75 | |
| M67465.1 | - | 37 | 75 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC092571.1, M67465.1, NM_012584.1, XM_003749360.3 transcripts.
Set Membership: |
|

LOC100911116
Genomic Map