Assay Details
| Gene Symbol: Abcb1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012623.2 | NP_036755.2 | 5 - 6 | 365 | 66 | |
| XM_008762687.1 | XP_008760909.1 | 4 - 5 | 503 | 66 | ||
| XM_008762688.1 | XP_008760910.1 | 4 - 5 | 502 | 66 | ||
| XM_008762691.1 | XP_008760913.1 | 4 - 5 | 497 | 66 | ||
| GenBank mRNA | AY082609.1 | - | 5 - 6 | 365 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abcb1b
Genomic Map