Assay Details
| Gene Symbol: Ppy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012626.2 | NP_036758.1 | 1 - 2 | 56 | 71 | |
| GenBank mRNA | M13588.1 | - | 1 - 2 | 56 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the M13588.1, NM_012626.2 transcripts.
Set Membership: |
|

Ppy
Genomic Map