Assay Details
| Gene Symbol: Psmb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012708.2 | NP_036840.2 | 1 - 2 | 113 | 95 | |
| GenBank mRNA | BC091161.1 | - | 1 - 2 | 101 | 7366097 | 95 |
| BP500440.1 | - | 1 - 2 | 118 | 95 | ||
| D10757.1 | - | 1 - 2 | 80 | 95 | ||
| FQ228039.1 | - | 1 - 2 | 87 | 95 | ||
| KC222887.1 | - | 1 - 2 | 62 | 95 | ||
| KC222888.1 | - | 1 - 2 | 62 | 95 | ||
| KC222889.1 | - | 1 - 2 | 62 | 95 | ||
| KC222891.1 | - | 1 - 2 | 62 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psmb9
Genomic Map