Assay Details
| Gene Symbol: Adra2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012739.3 | NP_036871.3 | 1 | 1006 | 123 | |
| GenBank mRNA | U79031.1 | - | 1 | 961 | 123 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Adra2a
Genomic Map