Assay Details
| Gene Symbol: Foxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012743.1 | NP_036875.1 | 1 - 2 | 189 | 93 | |
| GenBank mRNA | L09647.1 | - | 1 - 2 | 189 | 93 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the L09647.1,NM_012743.1 transcripts.
Set Membership: |
|

Foxa2
Genomic Map