Assay Details
| Gene Symbol: Syk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012758.1 | NP_036890.1 | 4 - 5 | 897 | 68 | |
| GenBank mRNA | L20838.1 | - | 2 - 3 | 533 | 68 | |
| U21683.1 | - | 4 - 5 | 897 | 68 | ||
| U21684.1 | - | 4 - 5 | 897 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
angiogenesis
serotonin secretion
lymph vessel development
adaptive immune response
macrophage activation involved in immune response
neutrophil activation involved in immune response
leukocyte activation involved in immune response
serotonin secretion by platelet
protein phosphorylation
inflammatory response
leukocyte cell-cell adhesion
cell surface receptor signaling pathway
enzyme linked receptor protein signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
activation of JUN kinase activity
regulation of platelet activation
cell migration
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
leukotriene biosynthetic process
signal transduction by protein phosphorylation
neutrophil chemotaxis
regulation of superoxide anion generation
positive regulation of cell adhesion mediated by integrin
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
defense response to bacterium
transcription factor import into nucleus
positive regulation of mast cell degranulation
regulation of neutrophil degranulation
beta selection
innate immune response
positive regulation of interleukin-3 biosynthetic process
positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
positive regulation of B cell differentiation
positive regulation of gamma-delta T cell differentiation
positive regulation of bone resorption
positive regulation of alpha-beta T cell differentiation
positive regulation of alpha-beta T cell proliferation
protein autophosphorylation
blood vessel morphogenesis
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
regulation of phagocytosis
regulation of immune response
positive regulation of calcium-mediated signaling
B cell receptor signaling pathway
regulation of sequence-specific DNA binding transcription factor activity
regulation of ERK1 and ERK2 cascade
cellular response to molecule of fungal origin
regulation of arachidonic acid secretion
regulation of platelet aggregation
protein kinase activity
protein serine/threonine kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
receptor binding
integrin binding
protein binding
ATP binding
protein kinase binding
protein domain specific binding
activation of MAPK activity
angiogenesis
serotonin secretion
lymph vessel development
adaptive immune response
macrophage activation involved in immune response
neutrophil activation involved in immune response
leukocyte activation involved in immune response
serotonin secretion by platelet
protein phosphorylation
inflammatory response
leukocyte cell-cell adhesion
cell surface receptor signaling pathway
enzyme linked receptor protein signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
activation of JUN kinase activity
regulation of platelet activation
cell migration
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
leukotriene biosynthetic process
signal transduction by protein phosphorylation
neutrophil chemotaxis
regulation of superoxide anion generation
positive regulation of cell adhesion mediated by integrin
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
defense response to bacterium
transcription factor import into nucleus
positive regulation of mast cell degranulation
regulation of neutrophil degranulation
beta selection
innate immune response
positive regulation of interleukin-3 biosynthetic process
positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
positive regulation of B cell differentiation
positive regulation of gamma-delta T cell differentiation
positive regulation of bone resorption
positive regulation of alpha-beta T cell differentiation
positive regulation of alpha-beta T cell proliferation
protein autophosphorylation
blood vessel morphogenesis
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
regulation of phagocytosis
regulation of immune response
positive regulation of calcium-mediated signaling
B cell receptor signaling pathway
regulation of sequence-specific DNA binding transcription factor activity
regulation of ERK1 and ERK2 cascade
cellular response to molecule of fungal origin
regulation of arachidonic acid secretion
regulation of platelet aggregation
protein kinase activity
protein serine/threonine kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
receptor binding
integrin binding
protein binding
ATP binding
protein kinase binding
protein domain specific binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017328 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017329 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P293904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509785 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285754 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135958 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150689 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017328 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017329 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P293904 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P293904 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017328 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017329 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P293904 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017329 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017328 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P293904 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23178 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46709 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39635 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48766 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15437 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6300 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10797525 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797514 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797517 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797513 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797518 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797516 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797520 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797512 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797511 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797515 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797524 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797521 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797519 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10797509 | - | - |
| Rat Genome 230 2.0 Array | 1368186_a_at | - | - |
| Rat Genome 230 2.0 Array | 1368186_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368186_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368186_PM_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700009663.rn.1 | TC1700001381.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1700009662.rn.1 | TC1700001381.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368186_a_at | - | - |
| Rat Expression Set 230 | 1368186_PM_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10797509 | - | - |
| Rat Genome U34 Set | L20838_s_at | - | - |
| Rat Genome U34 Set | U21683_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22029786 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350279 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65291 | - | - |

Syk
Genomic Map