Assay Details
| Gene Symbol: Cnr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012784.4 | NP_036916.1 | 1 - 2 | 89 | 81 | |
| GenBank mRNA | X55812.1 | - | 1 - 2 | 89 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_012784.4, X55812.1 transcripts.
Set Membership: |
|

Cnr1
Genomic Map