Assay Details
| Gene Symbol: Gstt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012796.2 | NP_036928.1 | 2 - 3 | 336 | 67 | |
| GenBank mRNA | BC061856.1 | - | 2 - 3 | 340 | 5623709 | 67 |
| CK473092.1 | - | 2 - 3 | 336 | 67 | ||
| D10026.1 | - | 2 - 3 | 324 | 67 | ||
| FJ179409.1 | - | 2 - 3 | 202 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gstt2
Genomic Map