Assay Details
| Gene Symbol: Acsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012820.1 | NP_036952.1 | 17 - 18 | 1799 | 74 | |
| XM_006253122.1 | XP_006253184.1 | 18 - 19 | 2459 | 74 | ||
| XM_006253123.1 | XP_006253185.1 | 18 - 19 | 2000 | 74 | ||
| XM_006253124.2 | XP_006253186.1 | 18 - 19 | 1919 | 74 | ||
| XM_006253125.2 | XP_006253187.1 | 18 - 19 | 2079 | 74 | ||
| XM_006253126.1 | XP_006253188.1 | 18 - 19 | 1925 | 74 | ||
| XM_006253127.1 | XP_006253189.1 | 18 - 19 | 1890 | 74 | ||
| XM_008771254.1 | XP_008769476.1 | 17 - 18 | 1825 | 74 | ||
| GenBank mRNA | D90109.1 | - | 17 - 18 | 1799 | 74 | |
| FQ217275.1 | - | 17 - 18 | 1769 | 74 | ||
| FQ219468.1 | - | 1 - 2 | 100 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Acsl1
Genomic Map