Assay Details
| Gene Symbol: Ephx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034090.3 | NP_001029262.1 | 3 - 4 | 504 | 70 | |
| NM_012844.3 | NP_036976.2 | 3 - 4 | 533 | 70 | ||
| GenBank mRNA | BC061568.1 | - | 3 - 4 | 464 | 6889368 | 70 |
| M26125.1 | - | 509 | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ephx1
Genomic Map