Assay Details
| Gene Symbol: Htr1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012852.1 | NP_036984.1 | 1 | 548 | 101 | |
| XM_008764208.1 | XP_008762430.1 | 2 | 1341 | 101 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Htr1d
Genomic Map