Assay Details
| Gene Symbol: Sstr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012882.2 | NP_037014.2 | 1 - 2 | 120 | 110 | |
| GenBank mRNA | L04535.1 | - | 1 - 2 | 120 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the L04535.1, NM_012882.2 transcripts.
Set Membership: |
|

Sstr5
Genomic Map