Assay Details
| Gene Symbol: Cnga2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012928.1 | NP_037060.1 | 321 | 97 | ||
| GenBank mRNA | AF126808.1 | - | 215 | 97 | ||
| X55519.1 | - | 321 | 97 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF126808.1, NM_012928.1, X55519.1 transcripts.
Set Membership: |
|

Cnga2
Genomic Map