Assay Details
| Gene Symbol: Pcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC059147.1 | - | 2 - 3 | 114 | 6888437 | 127 |
| BG665749.1 | - | 2 - 3 | 113 | 127 | ||
| BG666168.1 | - | 2 - 3 | 104 | 127 | ||
| CB613293.1 | - | 2 - 3 | 109 | 127 | ||
| FQ097405.1 | - | 1 - 2 | 127 | |||
| M24852.1 | - | 2 - 3 | 101 | 127 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC059147.1, M24852.1 transcripts.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P149562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179474 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397736 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57837 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56570 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P149562 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P149562 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P149562 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P346637 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68836 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10476 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99127 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22545 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11091 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700249996 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10750510 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750507 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750509 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750506 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750508 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10750505 | - | - |
| Rat Genome 230 2.0 Array | 1368145_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368145_at | - | - |
| Affymetrix HT Rat Focus Array | 1368145_at | - | - |
| Affymetrix HT Rat Focus Array | 1368145_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100001785.rn.1 | TC1100000361.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368145_PM_at | - | - |
| Rat Expression Set 230 | 1368145_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10750505 | - | - |
| Rat Genome U34 Set | M24852_at | - | - |
| Rat Genome U34 Set | M24852_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21330773 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357041 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63874 | - | - |

Pcp4
Genomic Map