Assay Details
| Gene Symbol: Gckr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013120.2 | NP_037252.1 | 8 - 9 | 822 | 67 | |
| GenBank mRNA | BC081843.1 | - | 8 - 9 | 822 | 7096237 | 67 |
| KJ026952.1 | - | 8 - 9 | 645 | 67 | ||
| X68497.1 | - | 8 - 9 | 666 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gckr
Genomic Map