Assay Details
| Gene Symbol: Dgkg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013126.1 | NP_037258.1 | 7 - 8 | 894 | 112 | |
| XM_006248538.2 | XP_006248600.1 | 7 - 8 | 1187 | 112 | ||
| XM_006248539.2 | XP_006248601.1 | 7 - 8 | 1186 | 112 | ||
| XM_006248540.2 | XP_006248602.1 | 7 - 8 | 1194 | 112 | ||
| GenBank mRNA | D38448.1 | - | 7 - 8 | 894 | 112 | |
| FQ231209.1 | - | 7 - 8 | 900 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P179145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296496 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150417 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719345 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823348 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 153588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111403 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132048 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P179145 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P179145 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P179145 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P179145 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700251638 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700494986 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99357 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33700 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33017 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51319 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36744 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89757 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78140 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11601 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14179 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102326 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73826 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86016 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 83489 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89759 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67641 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16546 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700251638 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700494986 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10752023 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752028 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752027 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752025 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752022 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752017 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752019 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752032 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752026 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752029 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752018 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752012 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752033 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752015 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752016 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752020 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752013 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752031 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752030 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752024 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752021 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10752007 | - | - |
| Rat Genome 230 2.0 Array | 1387265_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387265_at | - | - |
| Affymetrix HT Rat Focus Array | 1387265_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387265_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1100004952.rn.1 | TC1100000891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100004949.rn.1 | TC1100000891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100004950.rn.1 | TC1100000891.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1100004951.rn.1 | TC1100000891.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387265_at | - | - |
| Rat Expression Set 230 | 1387265_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10752007 | - | - |
| Rat Genome U34 Set | rc_AI233421_s_at | - | - |
| Rat Genome U34 Set | D38448_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21222484 | - | - |

Dgkg
Genomic Map