Assay Details
| Gene Symbol: Eif2ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013223.1 | NP_037355.1 | 3 - 4 | 425 | 75 | |
| GenBank mRNA | BC081838.1 | - | 3 - 4 | 458 | 7132671 | 75 |
| L27707.1 | - | 3 - 4 | 425 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
acute inflammatory response
translational initiation
regulation of translation
protein phosphorylation
phagocytosis
negative regulation of cell proliferation
regulation of translational initiation by eIF2 alpha phosphorylation
regulation of eIF2 alpha phosphorylation by heme
macrophage differentiation
negative regulation of translational initiation
protoporphyrinogen IX metabolic process
protein autophosphorylation
regulation of hemoglobin biosynthetic process
negative regulation of hemoglobin biosynthetic process
iron ion homeostasis
protein kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
ATP binding
heme binding
protein homodimerization activity
acute inflammatory response
translational initiation
regulation of translation
protein phosphorylation
phagocytosis
negative regulation of cell proliferation
regulation of translational initiation by eIF2 alpha phosphorylation
regulation of eIF2 alpha phosphorylation by heme
macrophage differentiation
negative regulation of translational initiation
protoporphyrinogen IX metabolic process
protein autophosphorylation
regulation of hemoglobin biosynthetic process
negative regulation of hemoglobin biosynthetic process
iron ion homeostasis
protein kinase activity
eukaryotic translation initiation factor 2alpha kinase activity
ATP binding
heme binding
protein homodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P112354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220854 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702322 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79844 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155422 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37452 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P112354 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P112354 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P112354 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P841516 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700324422 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 36584 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92638 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9252 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20322 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700324422 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10760371 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760374 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760363 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760368 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760373 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760362 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760361 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760360 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760372 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760364 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760367 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760366 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760359 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760369 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760370 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760365 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10760358 | - | - |
| Rat Genome 230 2.0 Array | 1374402_at | - | - |
| Rat Genome 230 2.0 Array | 1374402_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387272_at | - | - |
| Rat Genome 230 2.0 Array | 1387272_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387272_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387272_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200009535.rn.1 | TC1200001110.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387272_PM_at | - | - |
| Rat Expression Set 230 | 1387272_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10760358 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21718668 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363072 | - | - |
| RatRef-12 Expression BeadChip | ILMN_50048 | - | - |

Eif2ak1
Genomic Map