Assay Details
| Gene Symbol: Pde4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017031.2 | NP_058727.2 | 4 - 5 | 1056 | 70 | |
| GenBank mRNA | AF202732.1 | - | 914 | 70 | ||
| AF202733.1 | - | 4 - 5 | 620 | 70 | ||
| J04563.1 | - | 1 - 2 | 68 | 70 | ||
| U95748.1 | - | 4 - 5 | 1056 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pde4b
Genomic Map