Assay Details
| Gene Symbol: Ppp2cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017040.1 | NP_058736.1 | 2 - 3 | 622 | 81 | |
| GenBank mRNA | BC085926.1 | - | 2 - 3 | 606 | 7109733 | 81 |
| M23591.1 | - | 2 - 3 | 622 | 81 | ||
| X14087.1 | - | 2 - 3 | 455 | 81 | ||
| X16044.1 | - | 2 - 3 | 357 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2cb
Genomic Map