Assay Details
| Gene Symbol: Lox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017061.2 | NP_058757.1 | 5 - 6 | 1181 | 96 | |
| XM_006254713.2 | XP_006254775.1 | 3 - 4 | 1264 | 96 | ||
| XM_006254714.2 | XP_006254776.1 | 4 - 5 | 1164 | 96 | ||
| XM_006254715.2 | XP_006254777.1 | 3 - 4 | 1265 | 96 | ||
| GenBank mRNA | BC078861.1 | - | 5 - 6 | 1181 | 7113042 | 96 |
| FQ230374.1 | - | 3 - 4 | 416 | 96 | ||
| FQ231737.1 | - | 2 - 3 | 205 | 96 | ||
| U11038.1 | - | 3 - 4 | 1149 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lox
Genomic Map