Assay Details
| Gene Symbol: Chm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017067.1 | NP_058763.1 | 14 - 15 | 1771 | 68 | |
| GenBank mRNA | L13722.1 | - | 14 - 15 | 1771 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P071664 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P071659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662301 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662599 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739186 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970996 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158757 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116410 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450868 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4784 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114087 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071659 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071664 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071659 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P071664 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P071659 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P071664 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P472828 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17301 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98939 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93476 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71917 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47269 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3183 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 45009 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 15390 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59613 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 21277 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84624 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53758 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1716 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10939149 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939156 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939150 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939153 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939157 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939158 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939154 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939148 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939155 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939160 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939159 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939152 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939162 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939161 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939151 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10939147 | - | - |
| Rat Genome 230 2.0 Array | 1373802_at | - | - |
| Rat Genome 230 2.0 Array | 1369227_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373802_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369227_at | - | - |
| Affymetrix HT Rat Focus Array | 1373802_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369227_at | - | - |
| Affymetrix HT Rat Focus Array | 1373802_at | - | - |
| Affymetrix HT Rat Focus Array | 1369227_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00013377.rn.1 | TC0X00002102.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00013376.rn.1 | TC0X00002102.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00013380.rn.1 | TC0X00002102.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369227_at | - | - |
| Rat Expression Set 230 | 1373802_PM_at | - | - |
| Rat Expression Set 230 | 1369227_PM_at | - | - |
| Rat Expression Set 230 | 1373802_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939147 | - | - |
| Rat Genome U34 Set | L13722_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21374083 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_2039780 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62740 | - | - |

Chm
Genomic Map