Assay Details
| Gene Symbol: Stmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017166.1 | NP_058862.1 | 1 - 2 | 109 | 74 | |
| GenBank mRNA | J04979.1 | - | 1 - 2 | 109 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the J04979.1, NM_017166.1 transcripts.
Set Membership: |
|

Stmn1
Genomic Map