Assay Details
Gene Symbol: Gap43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017195.3 | NP_058891.1 | 1 - 2 | 252 | 83 | |
GenBank mRNA | BC127502.1 | - | 1 - 2 | 100 | 8364847 | 83 |
FQ214107.1 | - | 1 - 2 | 87 | 83 | ||
M16228.1 | - | 1 - 2 | 105 | 83 | ||
M16736.1 | - | 1 - 2 | 88 | 83 | ||
X06338.1 | - | 1 - 2 | 252 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC127502.1, M16228.1, M16736.1, NM_017195.3, X06338.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
