Assay Details
Gene Symbol: Map2k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033987.1 | NP_001029159.1 | 4 - 5 | 922 | 68 | |
NM_017246.2 | NP_058942.1 | 4 - 5 | 922 | 68 | ||
XM_006243229.2 | XP_006243291.1 | 4 - 5 | 981 | 68 | ||
XM_008766221.1 | XP_008764443.1 | 2 - 3 | 120 | 68 | ||
GenBank mRNA | BC078860.1 | - | 4 - 5 | 922 | 7122708 | 68 |
U37462.1 | - | 4 - 5 | 410 | 68 | ||
U37463.1 | - | 2 - 3 | 131 | 68 | ||
U37464.1 | - | 4 - 5 | 410 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
activation of MAPK activity
protein phosphorylation
heart development
peptidyl-tyrosine phosphorylation
positive regulation of cell growth
negative regulation of NF-kappaB transcription factor activity
negative regulation of heterotypic cell-cell adhesion
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of interleukin-8 biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of epithelial cell proliferation
positive regulation of protein metabolic process
negative regulation of response to cytokine stimulus
ERK5 cascade
cellular response to growth factor stimulus
negative regulation of cell migration involved in sprouting angiogenesis
negative regulation of chemokine (C-X-C motif) ligand 2 production
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
protein tyrosine kinase activity
ATP binding
metal ion binding
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
activation of MAPK activity
protein phosphorylation
heart development
peptidyl-tyrosine phosphorylation
positive regulation of cell growth
negative regulation of NF-kappaB transcription factor activity
negative regulation of heterotypic cell-cell adhesion
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of interleukin-8 biosynthetic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of epithelial cell proliferation
positive regulation of protein metabolic process
negative regulation of response to cytokine stimulus
ERK5 cascade
cellular response to growth factor stimulus
negative regulation of cell migration involved in sprouting angiogenesis
negative regulation of chemokine (C-X-C motif) ligand 2 production
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase activity
protein tyrosine kinase activity
ATP binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P485589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634418 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461888 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625999 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402805 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554037 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415749 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814840 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 391011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811069 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421596 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576171 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851411 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564353 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30367 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696717 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102283 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47806 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P485589 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P485589 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P485589 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P485589 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700590652 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10810 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72178 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 75610 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94263 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31705 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3035 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79305 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48973 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31526 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8632 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 77787 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86423 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88412 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66706 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97600 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50177 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 60489 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97848 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700590652 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10918128 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918135 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918141 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918140 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918142 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918129 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918130 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918136 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918127 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918132 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918138 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918131 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918133 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918126 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918139 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918134 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918137 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918120 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10918118 | - | - |
Rat Genome 230 2.0 Array | 1386959_a_at | - | - |
Rat Genome 230 2.0 Array | 1386959_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1386959_a_at | - | - |
Affymetrix HT Rat Focus Array | 1386959_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0800022180.rn.1 | TC0800002730.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0800022179.rn.1 | TC0800002730.rn.1 | RTA TC |
Rat Expression Set 230 | 1386959_PM_a_at | - | - |
Rat Expression Set 230 | 1386959_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10918118 | - | - |
Rat Genome U34 Set | U37464_s_at | - | - |
Rat Genome U34 Set | U37462_s_at | - | - |
Rat Genome U34 Set | U37464_s_at | - | - |
Rat Genome U34 Set | U37462_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21351778 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376514 | - | - |
