Assay Details
| Gene Symbol: Htr2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017254.1 | NP_058950.1 | 1 - 2 | 475 | 71 | |
| GenBank mRNA | M30705.1 | - | 1 - 2 | 475 | 71 | |
| M64867.1 | - | 881 | 71 | |||
| X13971.1 | - | 1018 | 71 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Htr2a
Genomic Map