Assay Details
| Gene Symbol: Gpam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017274.1 | NP_058970.1 | 10 - 11 | 1264 | 87 | |
| XM_006231625.2 | XP_006231687.1 | 10 - 11 | 1655 | 87 | ||
| XM_006231626.2 | XP_006231688.1 | 11 - 12 | 1536 | 87 | ||
| XM_008760534.1 | XP_008758756.1 | 10 - 11 | 1206 | 87 | ||
| GenBank mRNA | AF021348.1 | - | 10 - 11 | 1264 | 87 | |
| U36771.2 | - | 9 - 10 | 1040 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid metabolic process
acyl-CoA metabolic process
triglyceride metabolic process
glycerophospholipid metabolic process
phospholipid biosynthetic process
response to glucose
response to fructose
positive regulation of triglyceride biosynthetic process
response to activity
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
response to nutrient levels
cellular response to insulin stimulus
positive regulation of multicellular organism growth
positive regulation of activated T cell proliferation
response to cadmium ion
regulation of cytokine secretion
defense response to virus
fatty acid homeostasis
phospholipid homeostasis
negative regulation of activation-induced cell death of T cells
interleukin-2 secretion
glycerol-3-phosphate O-acyltransferase activity
fatty acid metabolic process
acyl-CoA metabolic process
triglyceride metabolic process
glycerophospholipid metabolic process
phospholipid biosynthetic process
response to glucose
response to fructose
positive regulation of triglyceride biosynthetic process
response to activity
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
response to nutrient levels
cellular response to insulin stimulus
positive regulation of multicellular organism growth
positive regulation of activated T cell proliferation
response to cadmium ion
regulation of cytokine secretion
defense response to virus
fatty acid homeostasis
phospholipid homeostasis
negative regulation of activation-induced cell death of T cells
interleukin-2 secretion
glycerol-3-phosphate O-acyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P242969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955849 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736488 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457122 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298354 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965803 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125689 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P242969 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P242969 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P242969 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P242969 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71853 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101318 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40417 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52503 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10730912 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730903 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730917 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730907 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730902 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730910 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730918 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730900 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730915 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730905 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730909 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730913 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730901 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730914 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730904 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730919 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730911 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730906 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730916 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730899 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730908 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730920 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10730898 | - | - |
| Rat Genome 230 2.0 Array | 1382986_at | - | - |
| Rat Genome 230 2.0 Array | 1382986_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369758_at | - | - |
| Rat Genome 230 2.0 Array | 1369758_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382986_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382986_at | - | - |
| Affymetrix HT Rat Focus Array | 1369758_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369758_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100063866.rn.1 | TC0100007927.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369758_PM_at | - | - |
| Rat Expression Set 230 | 1369758_at | - | - |
| Rat Expression Set 230 | 1382986_at | - | - |
| Rat Expression Set 230 | 1382986_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730898 | - | - |
| Rat Genome U34 Set | U36773_at | - | - |
| Rat Genome U34 Set | U36773_g_at | - | - |
| Rat Genome U34 Set | U36771_g_at | - | - |
| Rat Genome U34 Set | U36772_at | - | - |
| Rat Genome U34 Set | U36771_at | - | - |
| Rat Genome U34 Set | rc_AI146190_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22131826 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363492 | - | - |
| RatRef-12 Expression BeadChip | ILMN_68085 | - | - |

Gpam
Genomic Map