Assay Details
Gene Symbol: Atp2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110139.2 | NP_001103609.1 | 5 - 6 | 971 | 83 | |
NM_001110823.2 | NP_001104293.1 | 5 - 6 | 971 | 83 | ||
NR_027839.1 | 5 - 6 | 971 | 83 | |||
GenBank mRNA | J04022.1 | - | 5 - 6 | 969 | 83 | |
J04023.1 | - | 5 - 6 | 604 | 83 | ||
J04024.1 | - | 5 - 6 | 703 | 83 | ||
X15635.1 | - | 5 - 6 | 672 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
