Assay Details
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017327.1 | NP_059023.1 | 3 - 4 | 1012 | 63 | |
| XM_008772320.1 | XP_008770542.1 | 3 - 4 | 1021 | 63 | ||
| XM_008772321.1 | XP_008770543.1 | 3 - 4 | 1021 | 63 | ||
| XM_008772322.1 | XP_008770544.1 | 2 - 3 | 3630 | 63 | ||
| GenBank mRNA | M12671.1 | - | 2 - 3 | 176 | 63 | |
| M17526.1 | - | 3 - 4 | 1012 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
dopamine receptor signaling pathway
aging
locomotory behavior
regulation of heart contraction
metabolic process
cellular process
response to organonitrogen compound
response to organic cyclic compound
forebrain development
neuron projection development
response to cytokine
response to drug
response to hydrogen peroxide
response to morphine
positive regulation of GTPase activity
negative regulation of calcium ion transport
GTPase activity
signal transducer activity
protein binding
GTP binding
GDP binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
mu-type opioid receptor binding
protein complex binding
GTPase activating protein binding
metal ion binding
corticotropin-releasing hormone receptor 1 binding
G-protein coupled receptor signaling pathway
adenylate cyclase-modulating G-protein coupled receptor signaling pathway
dopamine receptor signaling pathway
aging
locomotory behavior
regulation of heart contraction
metabolic process
cellular process
response to organonitrogen compound
response to organic cyclic compound
forebrain development
neuron projection development
response to cytokine
response to drug
response to hydrogen peroxide
response to morphine
positive regulation of GTPase activity
negative regulation of calcium ion transport
GTPase activity
signal transducer activity
protein binding
GTP binding
GDP binding
G-protein beta/gamma-subunit complex binding
G-protein coupled serotonin receptor binding
mu-type opioid receptor binding
protein complex binding
GTPase activating protein binding
metal ion binding
corticotropin-releasing hormone receptor 1 binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P049836 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P115192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248081 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5243 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773940 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850158 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349479 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885318 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624610 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50520 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1986 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829527 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383741 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635435 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387163 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370344 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511763 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918304 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756171 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548903 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2258 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144592 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7288 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51104 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119839 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91047 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P149352 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P115192 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P149352 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P115192 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P149352 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P115192 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P489590 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P149352 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48472 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80816 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54123 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54106 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 859 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68199 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104153 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73361 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23174 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84984 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46332 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10809441 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809429 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809431 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809439 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809435 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809438 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809436 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809440 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809437 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809430 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809432 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10809428 | - | - |
| Rat Genome 230 2.0 Array | 1368879_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1368879_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368879_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368879_PM_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900008272.rn.1 | TC1900001026.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1900008271.rn.1 | TC1900001026.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368879_PM_a_at | - | - |
| Rat Expression Set 230 | 1368879_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809428 | - | - |
| Rat Genome U34 Set | M17526_g_at | - | - |
| Rat Genome U34 Set | M17526_at | - | - |
| Rat Genome U34 Set | rc_AI227704_at | - | - |
| Rat Genome U34 Set | M17526_at | - | - |
| Rat Genome U34 Set | M17526_g_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20879185 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650785 | - | - |
| RatRef-12 Expression BeadChip | ILMN_49964 | - | - |

Gnao1
Genomic Map