Assay Details
| Gene Symbol: Pdlim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017365.2 | NP_059061.1 | 1 - 2 | 165 | 96 | |
| GenBank mRNA | BC072465.1 | - | 1 - 2 | 156 | 7100446 | 96 |
| U23769.1 | - | 1 - 2 | 165 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P802550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82939 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8846 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871042 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118522 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P802550 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P802550 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P802550 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P802550 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 18386 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5608 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 23823 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700481935 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10730063 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730062 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730064 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730061 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730059 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10730057 | - | - |
| Rat Genome 230 2.0 Array | 1367946_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367946_at | - | - |
| Affymetrix HT Rat Focus Array | 1367946_at | - | - |
| Affymetrix HT Rat Focus Array | 1367946_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100061598.rn.1 | TC0100007673.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100061596.rn.1 | TC0100007673.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367946_at | - | - |
| Rat Expression Set 230 | 1367946_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730057 | - | - |
| Rat Genome U34 Set | U23769_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21345089 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361838 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66007 | - | - |

Pdlim1
Genomic Map