Assay Details
| Gene Symbol: Syn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110782.2 | NP_001104252.1 | 5 - 6 | 875 | 119 | |
| NM_019133.2 | NP_062006.1 | 5 - 6 | 875 | 119 | ||
| GenBank mRNA | M27812.1 | - | 5 - 6 | 857 | 119 | |
| M27924.1 | - | 5 - 6 | 857 | 119 | ||
| X04655.1 | - | 5 - 6 | 744 | 119 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P295251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100461 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925248 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162987 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P295251 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P295251 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P295251 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P262452 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89171 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58488 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 86979 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39388 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96523 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10932109 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932117 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932123 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932122 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932115 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932113 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932116 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932125 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932121 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932110 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932108 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932118 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932112 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932120 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932111 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932114 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932119 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10932107 | - | - |
| Rat Genome 230 2.0 Array | 1369251_a_at | - | - |
| Rat Genome 230 2.0 Array | 1390196_at | - | - |
| Rat Genome 230 2.0 Array | 1390196_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369251_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1369251_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1390196_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369251_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1390196_at | - | - |
| Rat Expression Set 230 | 1369251_a_at | - | - |
| Rat Expression Set 230 | 1369251_PM_a_at | - | - |
| Rat Expression Set 230 | 1390196_at | - | - |
| Rat Expression Set 230 | 1390196_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10932107 | - | - |
| Rat Genome U34 Set | M27812_at | - | - |
| Rat Genome U34 Set | rc_AI060071_at | - | - |
| Rat Genome U34 Set | M27812_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22234777 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373112 | - | - |
| RatRef-12 Expression BeadChip | ILMN_67983 | - | - |

Syn1
Genomic Map