Assay Details
| Gene Symbol: Tnfrsf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019135.1 | NP_062008.1 | 4 - 5 | 480 | 107 | |
| XM_008764206.1 | XP_008762428.1 | 4 - 5 | 1044 | 107 | ||
| GenBank mRNA | D42117.1 | - | 4 - 5 | 480 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
signal transduction
positive regulation of tumor necrosis factor biosynthetic process
positive regulation of apoptotic process
positive regulation of TRAIL biosynthetic process
positive regulation of NF-kappaB transcription factor activity
cellular response to mechanical stimulus
molecular_function
transmembrane signaling receptor activity
signal transduction
positive regulation of tumor necrosis factor biosynthetic process
positive regulation of apoptotic process
positive regulation of TRAIL biosynthetic process
positive regulation of NF-kappaB transcription factor activity
cellular response to mechanical stimulus
molecular_function
transmembrane signaling receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026661 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P013813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 695556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461851 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91292 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026661 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013813 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026661 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P013813 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P013813 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026661 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11943 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24946 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66346 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10881449 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881447 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881439 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881450 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881438 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881441 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881446 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881443 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881440 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881445 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881442 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881444 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881448 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10881437 | - | - |
| Rat Genome 230 2.0 Array | 1368635_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368635_at | - | - |
| Affymetrix HT Rat Focus Array | 1368635_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368635_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500030473.rn.1 | TC0500004012.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368635_at | - | - |
| Rat Expression Set 230 | 1368635_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10881437 | - | - |
| Rat Genome U34 Set | D42117_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21916973 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1651014 | - | - |

Tnfrsf8
Genomic Map