Assay Details
| Gene Symbol: Ugt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019276.3 | NP_062149.1 | 4 - 5 | 1117 | 97 | |
| GenBank mRNA | L21698.1 | - | 4 - 5 | 1164 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ugt8
Genomic Map