Assay Details
| Gene Symbol: Cym | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020091.1 | NP_064476.1 | 7 - 8 | 913 | 77 | |
| GenBank mRNA | AJ251688.1 | - | 7 - 8 | 913 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cym
Genomic Map