Assay Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020301.1 | NP_064697.1 | 16 - 17 | 1884 | 87 | |
| GenBank mRNA | X66140.1 | - | 16 - 17 | 1884 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P159300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33721 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220779 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343123 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792829 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156129 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72718 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P159300 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P159300 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P159300 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15500 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104914 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29141 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40111 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95775 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10784825 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784834 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784837 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784821 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784841 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784829 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784835 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784838 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784822 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784820 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784824 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784830 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784840 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784826 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784827 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784842 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784819 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784833 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784831 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784823 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784817 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784818 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784832 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784815 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784828 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784839 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10784814 | - | - |
| Rat Genome 230 2.0 Array | 1369823_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369823_at | - | - |
| Affymetrix HT Rat Focus Array | 1369823_at | - | - |
| Affymetrix HT Rat Focus Array | 1369823_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500013256.rn.1 | TC1500001952.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369823_at | - | - |
| Rat Expression Set 230 | 1369823_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10784814 | - | - |
| Rat Genome U34 Set | X66140_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21750146 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361457 | - | - |
| RatRef-12 Expression BeadChip | ILMN_57450 | - | - |

Adam7
Genomic Map