Assay Details
| Gene Symbol: Adam3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020302.1 | NP_064698.1 | 12 - 13 | 1355 | 77 | |
| GenBank mRNA | BC078839.1 | - | 12 - 13 | 1261 | 7114315 | 77 |
| Y07903.1 | - | 12 - 13 | 1355 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P351227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196447 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253195 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758800 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441343 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80733 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171896 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46125 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P351227 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P351227 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P351227 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P351227 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44473 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31109 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87409 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58671 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72064 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10788992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788976 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788972 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788973 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788980 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788990 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788977 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788988 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788984 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788983 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788986 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788985 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788971 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788989 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788970 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788987 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788974 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788975 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788979 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788978 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10788969 | - | - |
| Rat Genome 230 2.0 Array | 1369832_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369832_at | - | - |
| Affymetrix HT Rat Focus Array | 1369832_at | - | - |
| Affymetrix HT Rat Focus Array | 1369832_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600006438.rn.1 | TC1600000837.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600006439.rn.1 | TC1600000837.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600006437.rn.1 | TC1600000837.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369832_PM_at | - | - |
| Rat Expression Set 230 | 1369832_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10788969 | - | - |
| Rat Genome U34 Set | Y07903cds_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21560208 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367026 | - | - |
| RatRef-12 Expression BeadChip | ILMN_67200 | - | - |

Adam3a
Genomic Map