Assay Details
| Gene Symbol: LOC100911769 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762428.1 | XP_008760650.1 | 18 - 19 | 4222 | 81 | |
| XM_008762429.1 | XP_008760651.1 | 18 - 19 | 4219 | 81 | ||
| XM_008762430.1 | XP_008760652.1 | 18 - 19 | 7664 | 81 | ||
| XM_008762431.1 | XP_008760653.1 | 18 - 19 | 4198 | 81 | ||
| XM_008762432.1 | XP_008760654.1 | 17 - 18 | 4186 | 81 | ||
| XM_008762433.1 | XP_008760655.1 | 16 - 17 | 2305 | 81 | ||
| XM_008762434.1 | XP_008760656.1 | 15 - 16 | 2206 | 81 | ||
| XM_008762435.1 | XP_008760657.1 | 14 - 15 | 2170 | 81 | ||
| Gene Symbol: Epb41l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021681.2 | NP_067713.2 | 18 - 19 | 4252 | 81 | |
| NM_172090.3 | NP_742087.1 | 15 - 16 | 2219 | 81 | ||
| XM_008762381.1 | XP_008760603.1 | 18 - 19 | 4252 | 81 | ||
| GenBank mRNA | AB019256.1 | - | 15 - 16 | 2217 | 81 | |
| AB019257.1 | - | 18 - 19 | 4252 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100911769
Genomic Map