Assay Details
| Gene Symbol: Sik1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021693.2 | NP_067725.2 | 3 - 4 | 341 | 80 | |
| GenBank mRNA | AB020480.1 | - | 3 - 4 | 341 | 80 | |
| AF106937.1 | - | 399 | 80 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sik1
Genomic Map