Assay Details
| Gene Symbol: Nsf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021748.1 | NP_068516.1 | 950 | 61 | ||
| GenBank mRNA | AF142097.1 | - | 950 | 61 | ||
| AF189019.1 | - | 950 | 61 | |||
| FQ211783.1 | - | 989 | 61 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of receptor recycling
potassium ion transport
intra-Golgi vesicle-mediated transport
vesicle fusion
neurotransmitter secretion
metabolic process
protein transport
vesicle-mediated transport
regulation of exocytosis
Golgi to plasma membrane protein transport
Golgi vesicle docking
membrane fusion
protein binding
ATP binding
protein C-terminus binding
ATPase activity
syntaxin-1 binding
Rab GTPase binding
protein kinase binding
syntaxin binding
PDZ domain binding
D1 dopamine receptor binding
protein complex binding
ATPase activity, coupled
ATP-dependent protein binding
metal ion binding
positive regulation of receptor recycling
potassium ion transport
intra-Golgi vesicle-mediated transport
vesicle fusion
neurotransmitter secretion
metabolic process
protein transport
vesicle-mediated transport
regulation of exocytosis
Golgi to plasma membrane protein transport
Golgi vesicle docking
membrane fusion
protein binding
ATP binding
protein C-terminus binding
ATPase activity
syntaxin-1 binding
Rab GTPase binding
protein kinase binding
syntaxin binding
PDZ domain binding
D1 dopamine receptor binding
protein complex binding
ATPase activity, coupled
ATP-dependent protein binding
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P113493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270731 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582885 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 413402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279614 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296034 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960953 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631185 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 909153 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117205 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46312 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113493 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P113493 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P113493 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P243145 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99488 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 895 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44853 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26194 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40365 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34675 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78064 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5423 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69579 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104082 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10748120 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748138 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748126 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748119 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748132 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748127 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748124 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748136 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748135 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748123 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748129 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748131 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748137 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748122 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748139 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748133 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748130 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748134 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748128 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748121 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748125 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10748118 | - | - |
| Rat Genome 230 2.0 Array | 1369689_at | - | - |
| Rat Genome 230 2.0 Array | 1369689_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372528_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372528_at | - | - |
| Rat Genome 230 2.0 Array | 1369690_at | - | - |
| Rat Genome 230 2.0 Array | 1369690_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369689_at | - | - |
| Affymetrix HT Rat Focus Array | 1369690_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369690_at | - | - |
| Affymetrix HT Rat Focus Array | 1369689_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000035518.rn.1 | TC1000003911.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000035513.rn.1 | TC1000003911.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000035517.rn.1 | TC1000003911.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000035514.rn.1 | TC1000003911.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000035516.rn.1 | TC1000003911.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000035515.rn.1 | TC1000003911.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000035519.rn.1 | TC1000003911.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372528_PM_at | - | - |
| Rat Expression Set 230 | 1369690_at | - | - |
| Rat Expression Set 230 | 1369690_PM_at | - | - |
| Rat Expression Set 230 | 1372528_at | - | - |
| Rat Expression Set 230 | 1369689_PM_at | - | - |
| Rat Expression Set 230 | 1369689_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10748118 | - | - |
| Rat Genome U34 Set | AF091834_at | - | - |
| Rat Genome U34 Set | AF089839_at | - | - |
| Rat Genome U34 Set | rc_AI102069_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21223880 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363930 | - | - |
| RatRef-12 Expression BeadChip | ILMN_56089 | - | - |

Nsf
Genomic Map