Assay Details
| Gene Symbol: Il1rn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022194.2 | NP_071530.1 | 1 - 2 | 120 | 76 | |
| XM_006233636.2 | XP_006233698.1 | 2 - 3 | 2363 | 76 | ||
| XM_006233637.2 | XP_006233699.1 | 2 - 3 | 150 | 76 | ||
| XM_006233638.2 | XP_006233700.1 | 1 - 2 | 50 | 76 | ||
| GenBank mRNA | BC070930.1 | - | 1 - 2 | 114 | 76 | |
| CK364274.1 | - | 1 - 2 | 144 | 76 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006233637.2, XM_006233638.2 transcripts.
Set Membership: |
|

Il1rn
Genomic Map