Assay Details
| Gene Symbol: Necab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022302.1 | NP_071638.1 | 7 - 8 | 690 | 62 | |
| GenBank mRNA | AF193755.1 | - | 7 - 8 | 690 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P496624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95071 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333449 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 511025 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457824 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161276 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612491 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348457 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687386 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553419 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899293 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321636 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436550 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453103 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968946 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469702 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621997 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669215 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491651 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69221 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97040 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147083 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 142473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114960 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55212 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P496624 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P496624 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P496624 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P496624 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42300 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 1111 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26341 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85639 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 98954 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102617 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24823 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 71741 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 10836 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9737 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26071 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 25207 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 20029 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35499 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70444 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34710 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10875618 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875624 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875629 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875617 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875621 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875626 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875623 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875625 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875627 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875622 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875619 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875630 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875628 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875620 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10875616 | - | - |
| Rat Genome 230 2.0 Array | 1369540_at | - | - |
| Rat Genome 230 2.0 Array | 1369540_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369540_at | - | - |
| Affymetrix HT Rat Focus Array | 1369540_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500017748.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017756.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017742.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017747.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017750.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017752.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017741.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017749.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017753.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017746.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017740.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017743.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017745.rn.1 | TC0500002351.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500017755.rn.1 | TC0500002351.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369540_at | - | - |
| Rat Expression Set 230 | 1369540_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10875616 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21195844 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358305 | - | - |
| RatRef-12 Expression BeadChip | ILMN_51039 | - | - |

Necab1
Genomic Map