Assay Details
| Gene Symbol: Slc5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022590.2 | NP_072112.2 | 3 - 4 | 306 | 55 | |
| GenBank mRNA | AB070849.1 | - | 3 - 4 | 301 | 55 | |
| AB070850.1 | - | 3 - 4 | 301 | 55 | ||
| AB070976.1 | - | 3 - 4 | 301 | 55 | ||
| U29881.1 | - | 3 - 4 | 306 | 55 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a2
Genomic Map