Assay Details
| Gene Symbol: Pdap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022595.1 | NP_072117.1 | 1 - 2 | 37 | 59 | |
| GenBank mRNA | BC088140.1 | - | 1 - 2 | 51 | 7382664 | 59 |
| U26541.1 | - | 1 - 2 | 37 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC088140.1, NM_022595.1, U26541.1 transcripts.
Set Membership: |
|

Pdap1
Genomic Map